4-(Dimethylamino)piperidine ≥97%

Supplier: BeanTown Chemical

Synonyms: N,N-Dimethylpiperidin-4-amine

218665-25G 218665-5G
BT218665-25GEA 493 USD
BT218665-25G BT218665-5G
4-(Dimethylamino)piperidine ≥97%
4-(Dimethylamino)piperidine

CAS: 50533-97-6; EC No: 256-617-2; MDL No: MFCD00023144 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C7H16N2; MW: 128.22 Refractive Index: 1.476


  • Refractive index: 1.476
  • Liquid

Formula: C₇H₁₆N₂
MW: 128.219 g/mol
MDL Number: MFCD00023144
CAS Number: 50533-97-6
EINECS: 256-617-2
ADR: 8,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR