4-Fluoro-3-phenoxybenzaldehyde ≥98%

Supplier: BeanTown Chemical

218650-25G 218650-5G
BT218650-25GEA 209 USD
BT218650-25G BT218650-5G
4-Fluoro-3-phenoxybenzaldehyde ≥98%
4-Fluoro-3-phenoxybenzaldehyde

CAS: 68359-57-9; EC No: 269-856-2; MDL No: MFCD01318148 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: C13H9FO2; MW: 216.21 Density (g/mL): 1.2091; Refractive Index: 1.583


  • Refractive index: 1.583
  • Liquid

Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
Density: 1.2091 g/mL
MDL Number: MFCD01318148
CAS Number: 68359-57-9
EINECS: 269-856-2
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR