Isobutanol ≥99.0% ACS

Supplier: BeanTown Chemical

218595-1L 218595-4L 218595-500ML
BT218595-1LEA 66.69 USD
BT218595-1L BT218595-4L BT218595-500ML
Isobutanol ≥99.0% ACS
Isobutanol

CAS: 78-83-1; EC No: 201-148-0; MDL No: MFCD00004740; RTECS: NP9625000 UN No: UN1212; Haz Class: 3; Packing Group: III Liquid; Linear Formula: (CH3)CHCH2OH; Molecular Formula: C4H10O ; MW: 74.12 Melting Point: -108°; Boiling Point: 107-108°; Flash point: 28°C (82°F) Density (g/mL): 0.802; Refractive Index: 1.396


  • RTECS: NP9625000
  • Refractive index: 1.396
  • Liquid

Formula: (CH₃)₂CHCH₂OH
Boiling Pt: 107…108 °C
Melting Pt: –108 °C
Density: 0.802 g/mL
Flash Pt: 28 °C (82 °F)
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.
Color (APHA) ≤10
Residue after evaporation ≤10ppm
Solubility in water Passes test
Titrable acid ≤0.0005meq/g
Water ≤0.1%
Carbonyl compounds (as butyraldehyde) ≤0.01
2-Butanone ≤0.02%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR