Nickel(II) chloride, anhydrous ≥99% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Nickel dichloride

218515-100G 218515-500G
BT218515-100GEA 142.62 USD
BT218515-100G BT218515-500G
Nickel(II) chloride, anhydrous ≥99% (trace metals basis)
Nickel(II) chloride

CAS: 7718-54-9; EC No: 231-743-0; MDL No: MFCD00011142; RTECS: QR6475000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: NiCl2; MW: 129.60 Melting Point: 1001° Density (g/mL): 3.55 Hygroscopic


  • RTECS: QR6475000
  • Hygroscopic
  • Powder

Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1001 °C
Density: 3.55 g/mL
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR