Mesityl oxide ≥90% remainder 4-methyl-4-penten-2-one, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
4-Methyl-3-penten-2-one
218460-100ML
218460-500ML
BT218460-100MLEA
46
USD
BT218460-100ML
BT218460-500ML
Mesityl oxide ≥90% remainder 4-methyl-4-penten-2-one, Technical Grade
Mesityl oxide
CAS: 141-79-7; EC No: 205-502-5; MDL No: MFCD00008900; RTECS: SB4200000
UN No: UN1229; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: (CH3)2C=CHCOCH3; Molecular Formula: C6H10O; MW: 98.15
Melting Point: -41.5°; Boiling Point: 130°; Flash point: 30°C (86°F)
Density (g/mL): 0.858; Refractive Index: 1.442
- RTECS: SB4200000
- Refractive index: 1.442
- Liquid
Formula:
C₆H₁₀O MW: 98.14 g/mol Boiling Pt: 130 °C Melting Pt: –41.5 °C Density: 0.858 g/mL Flash Pt: 30 °C (86 °F) |
MDL Number:
MFCD00008900 CAS Number: 141-79-7 EINECS: 205-502-5 UN: 1229 ADR: 3,III |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...