Tantalum(V) isopropoxide 10% (w/v) in 2-propanol/hexane , 99.9% (trace metals basis)

Supplier: BeanTown Chemical

218455-100ML 218455-25ML
BT218455-100MLEA 416 USD
BT218455-100ML BT218455-25ML
Tantalum(V) isopropoxide 10% (w/v) in 2-propanol/hexane , 99.9% (trace metals basis)
Tantalum (V) isopropoxide

CAS: 16761-83-4; MDL No: MFCD00145347 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C15H35O5Ta; MW: 476.39 Moisture Sensitive


  • Moisture Sensitive
  • Liquid

Formula: C₁₅H₃₅O₅Ta
MW: 476.39 g/mol
MDL Number: MFCD00145347
CAS Number: 16761-83-4
ADR: 3,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR