(Chloromethyl)dimethylchlorosilane ≥98%

Supplier: BeanTown Chemical

Synonyms: CMDMCS, Chloro(chloromethyl)dimethylsilane

218435-100G 218435-25G
BT218435-100GEA 134 USD
BT218435-100G BT218435-25G
(Chloromethyl)dimethylchlorosilane ≥98%
(Chloromethyl)dimethylchlorosilane

CAS: 1719-57-9; EC No: 217-006-6; MDL No: MFCD00000875; RTECS: VV2095000 UN No: UN2985; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: ClCH2Si(CH3)2Cl; Molecular Formula: C3H8Cl2Si; MW: 143.09 Boiling Point: 114°/752 mmHg; Flash point: 22°C (72°F) Density (g/mL): 1.086; Refractive Index: 1.437 Moisture Sensitive


  • RTECS: VV2095000
  • Refractive index: 1.437
  • Moisture Sensitive
  • Liquid

Formula: C₃H₈Cl₂Si
MW: 143.09 g/mol
Boiling Pt: 114 °C/752 mmHg
Density: 1.086 g/mL
Flash Pt: 22 °C (72 °F)
MDL Number: MFCD00000875
CAS Number: 1719-57-9
EINECS: 217-006-6
UN: 2985
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR