Potassium tellurite hydrate ≥97%
Supplier: BeanTown Chemical
|
Danger
|
Synonyms:
Potassium tellurite(IV) hydrate, Potassium tellurium trioxide hydrate, di-Potassium tellurium trioxide hydrate, Potassium tellurium (IV) oxide hydrate, Potassium tellurite monohydrate
218365-100G
218365-25G
BT218365-100GEA
373.71
USD
BT218365-100G
BT218365-25G
Potassium tellurite hydrate ≥97%
Potassium tellurite hydrate
CAS: 123333-66-4; EC No: 232-213-1; MDL No: MFCD00149923
UN No: UN3284; Haz Class: 6.1; Packing Group: II
Crystalline; Linear Formula: K2TeO3·H2O ; MW: 276.81
Melting Point: 465° (decomposes)
Formula:
K₂O₃Te·H₂O MW: 271.81 g/mol Melting Pt: 465°C Dec. |
MDL Number:
MFCD00149923 CAS Number: 123333-66-4 EINECS: 232-213-1 UN: 3284 ADR: 6.1,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...