Sodium phenoxide ≥98%

Supplier: BeanTown Chemical

Synonyms: Sodium phenylate

218320-250G 218320-50G
BT218320-250GEA 287 USD
BT218320-250G BT218320-50G
Sodium phenoxide ≥98%
Sodium phenoxide

CAS: 139-02-6; EC No: 205-347-3; MDL No: MFCD00013134; RTECS: SM8780000 UN No: UN2905; Haz Class: 8; Packing Group: III Solid; Linear Formula: C6H5ONa; MW: 116.09 Melting Point: 300° Air Sensitive


  • RTECS: SM8780000
  • Air Sensitive
  • Solid

Formula: C₆H₅NaO
MW: 116.09 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00013134
CAS Number: 139-02-6
EINECS: 205-347-3
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR