Niobium ≥99.99% (trace metals basis), powder -325 mesh

Supplier: BeanTown Chemical

218315-10G 218315-250G 218315-50G
BT218315-10GEA 55.63 USD
BT218315-10G BT218315-250G BT218315-50G
Niobium ≥99.99% (trace metals basis), powder -325 mesh
Niobium

CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126; RTECS: QT9900000 UN No: UN3089; Haz Class: 4.1; Packing Group: II 10 Micron APS Powder; Molecular Formula: Nb ; MW: 92.91 Melting Point: 2468°; Boiling Point: 4742° Density (g/mL): 8.57


  • RTECS: QT9900000
  • 10 Micron APS Powder

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 4742 °C
Melting Pt: 2468 °C
Density: 8.57 g/mL
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR