Bromomethyl methyl ether ≥90%, Technical Grade
Supplier: BeanTown Chemical
218270-25G
218270-5G
BT218270-25GEA
178.23
USD
BT218270-25G
BT218270-5G
Bromomethyl methyl ether ≥90%, Technical Grade
Bromomethyl methyl ether
CAS: 13057-17-5; MDL No: MFCD00000171
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: II
Liquid; Linear Formula: BrCH2OCH3; Molecular Formula: C2H5BrO; MW: 124.96
Boiling Point: 87°; Flash point: 27°C (81°F)
Density (g/mL): 1.531; Refractive Index: 1.456
- Refractive index: 1.456
- Liquid
Formula:
C₂H₅BrO MW: 124.97 g/mol Boiling Pt: 87 °C Density: 1.531 g/mL Flash Pt: 27 °C (81 °F) |
MDL Number:
MFCD00000171 CAS Number: 13057-17-5 UN: 1992 ADR: 3 II |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...