Ergosterol hydrate ≥96% (dry weight)

Supplier: BeanTown Chemical

218230-25G 218230-5G
BT218230-25GEA 226.59 USD
BT218230-25G BT218230-5G
Ergosterol hydrate ≥96% (dry weight)
Ergosterol hydrate

CAS: 57-87-4; EC No: 200-352-7; MDL No: MFCD00003623 UN No: UN2811; Haz Class: 6.1; Packing Group: II Solid; Molecular Formula: C28H44O·xH2O; MW: 396.65 (anhydrous) Light Sensitive

Formula: C₂₈H₄₄O·xH₂O
MDL Number: MFCD00003623
CAS Number: 57-87-4
EINECS: 200-352-7
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR