3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,3-Bis(trifluoromethyl)-5-isothiocyanatobenzene, 1-Isothiocyanato-3,5-bis(trifluoromethyl)benzene
218195-1G
218195-25G
218195-5G
BT218195-1GEA
28
USD
BT218195-1G
BT218195-25G
BT218195-5G
3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate
CAS: 23165-29-9; MDL No: MFCD00040838
UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III
Liquid; Molecular Formula: C9H3F6NS; MW: 271.18
Boiling Point: 63°/1.5 mmHg; Flash point: 109°C (229°F)
Density (g/mL): 1.485; Refractive Index: 1.5
Moisture Sensitive
- Refractive index: 1.5
- Moisture Sensitive
- Liquid
Formula:
C₉H₃F₆NS MW: 271.19 g/mol Boiling Pt: 63 °C/1.5 mmHg Density: 1.485 g/mL Flash Pt: 109 °C (229 °F) |
MDL Number:
MFCD00040838 CAS Number: 23165-29-9 ADR: 8 (6.1),III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...