3,5-Dichloropyridine-2-carbonitrile ≥95%
Supplier: BeanTown Chemical
Synonyms:
3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine
218165-1G
218165-25G
218165-5G
BT218165-1GEA
35
USD
BT218165-1G
BT218165-25G
BT218165-5G
3,5-Dichloropyridine-2-carbonitrile ≥95%
3,5-Dichloropyridine-2-carbonitrile
CAS: 85331-33-5; MDL No: MFCD03788758
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H2Cl2N2; MW: 173.00
Melting Point: 99-103°
- Solid
Formula:
C₆H₂Cl₂N₂ Melting Pt: 99…103 °C |
MDL Number:
MFCD03788758 CAS Number: 85331-33-5 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...