Iron(II) chloride hydrate ≥99%

Supplier: BeanTown Chemical

218095-1KG 218095-250G
BT218095-1KGEA 139.86 USD
BT218095-1KG BT218095-250G
Iron(II) chloride hydrate ≥99%
Iron(II) chloride hydrate

CAS: 23838-02-0; EC No: 231-843-4; MDL No: MFCD00149711; RTECS: NO5600000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: FeCl2·xH2O; MW: 126.75 (anhydrous) Air Sensitive


  • RTECS: NO5600000
  • Air Sensitive
  • Crystalline

Formula: Cl₂Fe·xH₂O
MDL Number: MFCD00149711
CAS Number: 23838-02-0
EINECS: 231-843-4
ADR: 8,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR