4,4'-Methylenedianiline ≥97%
Supplier: BeanTown Chemical
Synonyms:
4,4’-Diaminodiphenylmethane, Bis(4-aminophenyl)methane, 4,4'-Diaminodiphenylmethane, 4,4'-Methylenedianiline
218090-1KG
218090-250G
218090-50G
BT218090-1KGEA
89.01
USD
BT218090-1KG
BT218090-250G
BT218090-50G
4,4'-Methylenedianiline ≥97%
4,4'-Methylenedianiline
CAS: 101-77-9; EC No: 202-974-4; MDL No: MFCD00007914; RTECS: BY5425000
UN No: UN2651; Haz Class: 6.1; Packing Group: III
Solid/Flakes; Molecular Formula: C13H14N2; MW: 198.26
Melting Point: 88-92°
Density (g/mL): 1.15
- RTECS: BY5425000
- Solid/Flakes
Formula:
C₁₃H₁₄N₂ MW: 198.27 g/mol Melting Pt: 88…92 °C Density: 1.15 g/mL |
MDL Number:
MFCD00007914 CAS Number: 101-77-9 EINECS: 202-974-4 UN: 2651 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...