Diindium triselenide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Indium (III) selenide, Indiumselenide
218060-25G
218060-5G
BT218060-25GEA
404
USD
BT218060-25G
BT218060-5G
Diindium triselenide ≥99.99%
Diindium triselenide
CAS: 12056-07-4; EC No: 235-016-9; MDL No: MFCD00016147
UN No: UN3283; Haz Class: 6.1; Packing Group: III
3-6mm Pieces; Molecular Formula: In2Se3; MW: 466.52
Density (g/mL): 5.67
- 3-6mm Pieces
Formula:
In₂Se₃ MW: 466.52 Density: 5.67 g/mL |
MDL Number:
MFCD00016147 CAS Number: 12056-07-4 EINECS: 235-016-9 UN: 3283 ADR: 6.1 III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...