4,5-Dichloro-o-phenylenediamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine
218045-100G
218045-25G
218045-5G
BT218045-100GEA
1018
USD
BT218045-100G
BT218045-25G
BT218045-5G
4,5-Dichloro-o-phenylenediamine ≥98%
4,5-Dichloro-o-phenylenediamine
CAS: 5348-42-5; EC No: 226-305-0; MDL No: MFCD00007723
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Molecular Formula: C6H6Cl2N2; MW: 177.03
Melting Point: 158-164°
- Crystalline/Powder
Formula:
C₆H₆Cl₂N₂ MW: 177.03 g/mol Melting Pt: 158…164 °C |
MDL Number:
MFCD00007723 CAS Number: 5348-42-5 EINECS: 226-305-0 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...