Potassium fluoride dihydrate ≥98.5%

Supplier: BeanTown Chemical

217980-1KG 217980-250G
BT217980-1KGEA 171 USD
BT217980-1KG BT217980-250G
Potassium fluoride dihydrate ≥98.5%
Potassium fluoride dihydrate

CAS: 13455-21-5; EC No: 232-151-5; MDL No: MFCD00149929 UN No: UN1812; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: KF·2H2O; MW: 94.13 Melting Point: 41°; Boiling Point: 156° Density (g/mL): 2.454 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: FK·2H₂O
MW: 94.13 g/mol
Boiling Pt: 156 °C
Melting Pt: 41 °C
Density: 2.454 g/mL
MDL Number: MFCD00149929
CAS Number: 13455-21-5
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR