Acrolein diethyl acetal ≥96%

Supplier: BeanTown Chemical

Synonyms: Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene

217895-10G 217895-50G
BT217895-10GEA 53 USD
BT217895-10G BT217895-50G
Acrolein diethyl acetal ≥96%
Acrolein diethyl acetal

CAS: 3054-95-3; EC No: 221-276-0; MDL No: MFCD00009239; RTECS: AS1370000 UN No: UN2374; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH2=CHCH(OCH2CH3)2 ; Molecular Formula: C7H14O2; MW: 130.18 Boiling Point: 125°; Flash point: 15°C (59°F) Density (g/mL): 0.854; Refractive Index: 1.398


  • RTECS: AS1370000
  • Refractive index: 1.398
  • Liquid

Formula: C₇H₁₄O₂
Boiling Pt: 125 °C
Density: 0.854 g/mL
Flash Pt: 15 °C (59 °F)
MDL Number: MFCD00009239
CAS Number: 3054-95-3
EINECS: 221-276-0
UN: 2374
ADR: 3,II

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR