Arsenic (III) sulfide ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Arsenic trisulphide, Disrsenic trisulphide
217890-25G
217890-5G
BT217890-25GEA
220
USD
BT217890-25G
BT217890-5G
Arsenic (III) sulfide ≥99.9% (trace metals basis)
Arsenic(III) sulphide
CAS: 1303-33-9; EC No: 215-117-4; MDL No: MFCD00003435; RTECS: CG2638000
UN No: UN1557; Haz Class: 6.1; Packing Group: III
1-6mm Fused Pieces; Molecular Formula: As2S3 ; MW: 246.03
- RTECS: CG2638000
- 1-6mm Fused Pieces
Formula:
As₂S₃ MW: 246.04 g/mol |
MDL Number:
MFCD00003435 CAS Number: 1303-33-9 EINECS: 215-117-4 UN: 1557 ADR: 6.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...