Nickel(II) stearate

Supplier: BeanTown Chemical

Synonyms: Nickel stearate, Nickel(II) stearate

217870-100G 217870-2KG 217870-500G
BT217870-100GEA 40 USD
BT217870-100G BT217870-2KG BT217870-500G
Nickel(II) stearate
Nickel(II) stearate

CAS: 2223-95-2; EC No: 218-744-1; MDL No: MFCD00054344 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: [CH3(CH2)16COO]2Ni; Molecular Formula: C36H70NiO4; MW: 625.63 Melting Point: 82-86° Density (g/mL): 1.13


  • Powder

Formula: C₃₆H₇₀NiO₄
MW: 625.632 g/mol
Melting Pt: 82…86 °C
Density: 1.13 g/mL
MDL Number: MFCD00054344
CAS Number: 2223-95-2
EINECS: 218-744-1
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR