1,3-Difluoroacetone ≥98%
Supplier: BeanTown Chemical
217750-1G
217750-5G
BT217750-1GEA
105
USD
BT217750-1G
BT217750-5G
1,3-Difluoroacetone ≥98%
1,3-Difluoroacetone
CAS: 453-14-5; MDL No: MFCD07368665
UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II
Liquid; Molecular Formula: C3H4F2O; MW: 94.06
Density (g/mL): 1.307; Refractive Index: 1.371
- Refractive index: 1.371
- Liquid
Formula:
C₃H₄F₂O MW: 94.06 g/mol Density: 1.307 g/mL |
MDL Number:
MFCD07368665 CAS Number: 453-14-5 UN: 2929 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...