1-Bromo-4-chlorobenzene ≥98%
Supplier: BeanTown Chemical
CAS: 106-39-8; EC No: 203-392-3; MDL No: MFCD00000600; RTECS: CY9006000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C6H4BrCl ; MW: 191.45
Melting Point: 64-67°; Boiling Point: 196°
Density (g/mL): 1.651
- RTECS: CY9006000
- Crystalline
Formula:
C₆H₄BrCl Boiling Pt: 196 °C Melting Pt: 64…67 °C Density: 1.651 g/mL |
MDL Number:
MFCD00000600 CAS Number: 106-39-8 EINECS: 203-392-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...