Lead zirconium titanium oxide (∼30 wt% Oxide) polymeric precursor
Supplier: BeanTown Chemical
Synonyms:
Lead zirconate titanate
217690-10G
217690-50G
BT217690-10GEA
92
USD
BT217690-10G
BT217690-50G
Lead zirconium titanium oxide (∼30 wt% Oxide) polymeric precursor
Lead zirconium titanium oxide
CAS: 12626-81-2; MDL No: MFCD00212270
UN No: UN1993; Haz Class: 3; Packing Group: III
Semi-Solid/Liquid
- Semi-Solid/Liquid
Formula:
PbZr(1-x)TixO₃ |
MDL Number:
MFCD00212270 CAS Number: 12626-81-2 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...