1,2,4-Trimethylbenzene ≥98%
Supplier: BeanTown Chemical
CAS: 95-63-6; EC No: 202-436-9; MDL No: MFCD00008527; RTECS: DC3325000
UN No: UN3295; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C9H12; MW: 120.19
Melting Point: -44°; Boiling Point: 168°; Flash point: 48°C (118°F)
Density (g/mL): 0.876; Refractive Index: 1.504
- RTECS: DC3325000
- Refractive index: 1.504
- Liquid
Formula:
C₉H₁₂ Boiling Pt: 168 °C Melting Pt: –44 °C Density: 0.876 g/mL Flash Pt: 48 °C (118 °F) |
MDL Number:
MFCD00008527 CAS Number: 95-63-6 EINECS: 202-436-9 UN: 3295 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...