Iron(III) nitrate nonahydrate 98.0-101.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
217660-2KG
217660-500G
BT217660-2KGEA
185.62
USD
BT217660-2KG
BT217660-500G
Iron(III) nitrate nonahydrate 98.0-101.0% ACS
Iron(III) nitrate nonahydrate
CAS: 7782-61-8; EC No: 233-899-5; MDL No: MFCD00149708; RTECS: NO7175000
UN No: UN1466; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Fe(NO3)3·9H2O; MW: 403.99
Melting Point: 47°; Boiling Point: 125° (decomposes)
Moisture Sensitive
- RTECS: NO7175000
- Moisture Sensitive
- Crystalline
Formula:
Fe(NO₃)₃·9H₂O MW: 404 g/mol Boiling Pt: 125 (dec.) Melting Pt: 47 |
MDL Number:
MFCD00149708 CAS Number: 7782-61-8 EINECS: 233-899-5 UN: 1466 ADR: 5.1 III Merck Index: 13,04057 |
Specification Test Results
Assay | 98.0-101.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...