Iron(III) ethoxide ≥99.5%
Supplier: BeanTown Chemical
217630-2G
217630-500MG
BT217630-10GEA
290.81
USD
BT217630-10G
BT217630-2G
BT217630-500MG
Iron(III) ethoxide ≥99.5%
Iron(III) ethoxide
CAS: 5058-42-4; EC No: 225-760-2; MDL No: MFCD00078028
UN No: UN3181; Haz Class: 4.1; Packing Group: III
Powder/Chunks; Linear Formula: Fe(OCH2CH3)3; MW: 191.03
Moisture Sensitive
- Moisture Sensitive
- Powder/Chunks
Formula:
C₆H₁₅FeO₃ MW: 191.03 g/mol |
MDL Number:
MFCD00078028 CAS Number: 5058-42-4 EINECS: 225-760-2 UN: 3181 ADR: 4.1 III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...