m-Cresol ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene

217615-1KG 217615-2.5KG 217615-250G
BT217615-1KGEA 76 USD
BT217615-1KG BT217615-2.5KG BT217615-250G
m-Cresol ≥99%
m-Cresol

CAS: 108-39-4; EC No: 203-577-9; MDL No: MFCD00002302; RTECS: GO6125000 UN No: UN2076; Haz Class: 6.1; Packing Group: II Liquid; Linear Formula: CH3C6H4OH; Molecular Formula: C7H8O; MW: 108.14 Melting Point: 8-10°; Boiling Point: 202-204°; Flash point: 86°C (187°F) Density (g/mL): 1.034; Refractive Index: 1.541


  • RTECS: GO6125000
  • Refractive index: 1.541
  • Liquid

Formula: CH₃C₆H₄OH
MW: 108.14 g/mol
Boiling Pt: 202…204 °C
Melting Pt: 8…10 °C
Density: 1.034 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00002302
CAS Number: 108-39-4
EINECS: 203-577-9
UN: 2076
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce465c84cc8b7c230526b22e6f830300b605d109&vid=72c08ae3da371936f9349196418a240331cb79ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR