Trimethylsilyl trifluoromethanesulphonate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Trimethylsilyl trifluoromethanesulphonate
217590-10G
217590-250G
217590-50G
BT217590-10GEA
46.2
USD
BT217590-10G
BT217590-250G
BT217590-50G
Trimethylsilyl trifluoromethanesulphonate ≥99%
Trimethylsilyl trifluoromethanesulphonate
CAS: 27607-77-8; EC No: 248-565-4; MDL No: MFCD00000406
UN No: UN2924; Haz Class: 3(8); Packing Group: III
Liquid; Linear Formula: (CH3)3SiSO3CF3; Molecular Formula: C4H9F3O3SSi; MW: 222.26
Boiling Point: 77°/80 mmHg; Flash point: 25°C (77°F)
Density (g/mL): 1.228; Refractive Index: 1.362
Moisture Sensitive
- Refractive index: 1.362
- Moisture Sensitive
- Liquid
Formula:
C₄H₉F₃O₃SSi MW: 222.26 g/mol Boiling Pt: 77 Density: 1.228 g/mL Flash Pt: 25 °C (77 °F) |
MDL Number:
MFCD00000406 CAS Number: 27607-77-8 EINECS: 248-565-4 ADR: 3(8) III Merck Index: 12,09849 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...