Ammonium persulfate (APS) ≥98%
Supplier: BeanTown Chemical
217575-1KG
217575-5KG
BT217575-1KGEA
81.49
USD
BT217575-1KG
BT217575-5KG
Ammonium persulfate (APS) ≥98%
Ammonium peroxodisulphate (APS)
CAS: 7727-54-0; EC No: 231-786-5; MDL No: MFCD00003390; RTECS: SE0350000
UN No: UN1444; Haz Class: 5.1; Packing Group: III
Powder; Linear Formula: (NH4)2S2O8; MW: 228.20
Moisture Sensitive
- RTECS: SE0350000
- Moisture Sensitive
- Powder
Formula:
(NH₄)₂S₂O₈ |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 EINECS: 231-786-5 UN: 1444 ADR: 5.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...