Oxazole ≥98%
Supplier: BeanTown Chemical
217535-1G
217535-5G
BT217535-1GEA
104.32
USD
BT217535-1G
BT217535-5G
Oxazole ≥98%
Oxazole
CAS: 288-42-6; EC No: 206-020-8; MDL No: MFCD00009751
UN No: UN2924; Haz Class: 3(8); Packing Group: II
Liquid; Molecular Formula: C3H3NO; MW: 69.07
Melting Point: -86°; Boiling Point: 69-70°; Flash point: 19°C (66°F)
Density (g/mL): 1.05; Refractive Index: 1.425
- Refractive index: 1.425
- Liquid
Formula:
C₃H₃NO MW: 69.06 g/mol Boiling Pt: 69…70 °C Melting Pt: –86 °C Density: 1.05 g/mL Flash Pt: 19 °C (66 °F) |
MDL Number:
MFCD00009751 CAS Number: 288-42-6 EINECS: 206-020-8 UN: 2924 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...