Mercury(II) trifluoroacetate ≥98%
Supplier: BeanTown Chemical
217515-25G
217515-5G
BT217515-25GEA
201
USD
BT217515-25G
BT217515-5G
Mercury(II) trifluoroacetate ≥98%
Mercury(II) trifluoroacetate
CAS: 13257-51-7; EC No: 236-250-4; MDL No: MFCD00013201
UN No: UN2025; Haz Class: 6.1; Packing Group: II
Crystalline/Powder; Linear Formula: Hg(OOCCF3)2; Molecular Formula: C4F6HgO4 ; MW: 426.62
Melting Point: 171-173°
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
C₄F₆HgO₄ MW: 426.62 g/mol Melting Pt: 171…173 °C |
MDL Number:
MFCD00013201 CAS Number: 13257-51-7 EINECS: 236-250-4 UN: 2025 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...