Bis(acetonitrile)palladium(II) dichloride
Supplier: BeanTown Chemical
Synonyms:
Bis(acetonitrile)-palladium(II) chloride, Palladium(II) chloride diacetonitrile complex, Dichlorobis(acetonitrile)palladium(II)
217505-1G
217505-5G
BT217505-1GEA
96
USD
BT217505-1G
BT217505-5G
Bis(acetonitrile)palladium(II) dichloride
Bis(acetonitrile)palladium(II) dichloride
CAS: 14592-56-4; EC No: 238-637-3; MDL No: MFCD00013122
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: PdCl2(CH3CN)2; Molecular Formula: C4H6Cl2N2Pd; MW: 259.43
- Powder
Formula:
C₄H₆Cl₂N₂Pd MW: 259.43 g/mol |
MDL Number:
MFCD00013122 CAS Number: 14592-56-4 EINECS: 238-637-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...