4-Nitrobenzenesulfonyl chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Nitrobenzene-1-sulphonyl chloride
217485-100G
217485-25G
217485-500G
BT217485-100GEA
170
USD
BT217485-100G
BT217485-25G
BT217485-500G
4-Nitrobenzenesulfonyl chloride ≥97%
4-Nitrobenzenesulphonyl chloride
CAS: 98-74-8; EC No: 202-697-9; MDL No: MFCD00007445
UN No: UN3261; Haz Class: 8; Packing Group: II
Powder; Molecular Formula: C6H4ClNO4S; MW: 221.62
Melting Point: 66-70°
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
C₆H₄ClNO₄S MW: 221.62 g/mol Melting Pt: 66…70 °C |
MDL Number:
MFCD00007445 CAS Number: 98-74-8 EINECS: 202-697-9 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...