Iron(III) nitrate nonahydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
CAS: 7782-61-8; EC No: 233-899-5; MDL No: MFCD00149708; RTECS: NO7175000
UN No: UN1466; Haz Class: 5.1; Packing Group: III
Crystalline Aggregates; Linear Formula: Fe(NO3)3·xH2O; MW: 241.86 (anhydrous)
Melting Point: 47°; Boiling Point: 125° (decomposes)
Moisture Sensitive
- RTECS: NO7175000
- Moisture Sensitive
- Crystalline Aggregates
Formula:
Fe(NO₃)₃·9H₂O Boiling Pt: 125 °C (dec.) Melting Pt: 47 °C |
MDL Number:
MFCD00149708 CAS Number: 7782-61-8 EINECS: 233-899-5 UN: 1466 ADR: 5.1,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...