5-Bromo-o-anisidine ≥97%

Supplier: BeanTown Chemical

217435-25G 217435-5G
BT217435-25GEA 591 USD
BT217435-25G BT217435-5G
5-Bromo-o-anisidine ≥97%
5-Bromo-o-anisidine

CAS: 6358-77-6; MDL No: MFCD04037882 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C7H8BrNO; MW: 202.05 Melting Point: 94-98°


  • Solid

Formula: C₇H₈BrNO
Melting Pt: 94…98 °C
MDL Number: MFCD04037882
CAS Number: 6358-77-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR