Nickel(II) hydroxide
Supplier: BeanTown Chemical
217400-1KG
217400-250G
BT217400-1KGEA
257
USD
BT217400-1KG
BT217400-250G
Nickel(II) hydroxide
Nickel(II) hydroxide
CAS: 12054-48-7; EC No: 235-008-5; MDL No: MFCD00011140; RTECS: QR7040000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: Ni(OH)2; MW: 92.70
Melting Point: 200° (decomposes)
Density (g/mL): 4.1
- RTECS: QR7040000
- Powder
Formula:
H₂NiO₂ MW: 92.71 g/mol Melting Pt: 200 °C (dec.) Density: 4.1 g/mL |
MDL Number:
MFCD00011140 CAS Number: 12054-48-7 EINECS: 235-008-5 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...