Platinum 5% on carbon

Supplier: BeanTown Chemical

Synonyms: Platinium

217370-1G 217370-25G 217370-5G
BT217370-1GEA 26 USD
BT217370-1G BT217370-25G BT217370-5G
Platinum 5% on carbon
Platinum

CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 UN No: UN1325; Haz Class: 4.1; Packing Group: III Solid


  • RTECS: TP2160000
  • Solid

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
ADR: 4.1 III
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR