2-Phenylethanol ≥98%

Supplier: BeanTown Chemical

Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol

217285-1KG 217285-250G 217285-5KG
BT217285-1KGEA 110.77 USD
BT217285-1KG BT217285-250G BT217285-5KG
2-Phenylethanol ≥98%
2-Phenylethanol

CAS: 60-12-8; EC No: 200-456-2; MDL No: MFCD00002886; RTECS: SG7175000 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: C6H5CH2CH2OH; Molecular Formula: C8H10O; MW: 122.16 Melting Point: -27°; Boiling Point: 219-221°; Flash point: 102°C (215°F) Density (g/mL): 1.02; Refractive Index: 1.532


  • RTECS: SG7175000
  • Refractive index: 1.532
  • Liquid

Formula: C₆H₅CH₂CH₂OH
MW: 122.17 g/mol
Boiling Pt: 432
Melting Pt: –27 °C
Density: 1.02 g/mL
Flash Pt: 102 °C (215 °F)
MDL Number: MFCD00002886
CAS Number: 60-12-8
EINECS: 200-456-2
UN: 2810
ADR: 6.1 III
Merck Index: 12,07370

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR