Methyl ethyl ketone ≥99%
Supplier: BeanTown Chemical
Synonyms:
EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
217280-1L
217280-2.5L
217280-500ML
BT217280-1LEA
46.95
USD
BT217280-1L
BT217280-2.5L
BT217280-500ML
Methyl ethyl ketone ≥99%
Methyl ethyl ketone
CAS: 78-93-3; EC No: 201-159-0; MDL No: MFCD00011648; RTECS: EL6475000
UN No: UN1193; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C4H8O; MW: 72.11
Melting Point: -87°; Boiling Point: 80°; Flash point: -3°C (27°F)
Density (g/mL): 0.805; Refractive Index: 1.379
- RTECS: EL6475000
- Refractive index: 1.379
- Liquid
Formula:
C₂H₅COCH₃ MW: 72.11 g/mol Boiling Pt: 79.6 Melting Pt: -86 Density: 0.805 g/mL Flash Pt: –3 °C (27 °F) |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3 II Merck Index: 13,06097 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...