Gold trichloride
Supplier: BeanTown Chemical
Synonyms:
Gold(III) chloride, Gold (III) chloride
217265-1G
217265-250MG
217265-5G
BT217265-1GEA
179.65
USD
BT217265-1G
BT217265-250MG
BT217265-5G
Gold trichloride
Gold trichloride
CAS: 13453-07-1; EC No: 236-623-1; MDL No: MFCD00014172; RTECS: MD5420000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: AuCl3; MW: 303.33
Density (g/mL): 3.9
Hygroscopic
- RTECS: MD5420000
- Hygroscopic
- Crystalline
Formula:
AuCl₃ MW: 303.32 g/mol Density: 3.9 g/mL |
MDL Number:
MFCD00014172 CAS Number: 13453-07-1 EINECS: 236-623-1 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...