1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane ≥97%

Supplier: BeanTown Chemical

217140-1G 217140-25G 217140-5G
BT217140-1GEA 57 USD
BT217140-1G BT217140-25G BT217140-5G
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane ≥97%
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane

CAS: 69304-37-6; MDL No: MFCD00009655 UN No: UN1760; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C12H28Cl2OSi2; MW: 315.43 Boiling Point: 70°/0.5 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.986; Refractive Index: 1.454 Moisture Sensitive


  • Refractive index: 1.454
  • Moisture Sensitive
  • Liquid

Formula: C₁₂H₂₈Cl₂OSi₂
Boiling Pt: 70 °C/0.5 mmHg
Density: 0.986 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00009655
CAS Number: 69304-37-6
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR