Dibromomethane ≥99%

Supplier: BeanTown Chemical

Synonyms: Methylenbromide

217110-100G 217110-2.5KG 217110-500G
BT217110-100GEA 28.46 USD
BT217110-100G BT217110-2.5KG BT217110-500G
Dibromomethane ≥99%
Dibromomethane

CAS: 74-95-3; EC No: 200-824-2; MDL No: MFCD00000168; RTECS: PA7350000 UN No: UN2664; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: CH2Br2; MW: 173.83 Melting Point: -52°; Boiling Point: 96-98° Density (g/mL): 2.477; Refractive Index: 1.541


  • RTECS: PA7350000
  • Refractive index: 1.541
  • Liquid

Formula: Br₂CH₂
MW: 173.83 g/mol
Boiling Pt: 96…98 °C
Melting Pt: –52 °C
Density: 2.477 g/mL
MDL Number: MFCD00000168
CAS Number: 74-95-3
EINECS: 200-824-2
UN: 2664
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR