2,4-Difluorophenol ≥98%
Supplier: BeanTown Chemical
CAS: 367-27-1; EC No: 206-688-0; MDL No: MFCD00009715
UN No: UN2921; Haz Class: 8 (4.1); Packing Group: II
Liquid/Low Melting Solid; Molecular Formula: C6H4F2O; MW: 130.09
Melting Point: 22.4°; Boiling Point: 52-53°/19 mmHg; Flash point: 57°C (135°F)
Density (g/mL): 1.362; Refractive Index: 1.486
- Refractive index: 1.486
- Liquid/Low Melting Solid
Formula:
C₆H₄F₂O Boiling Pt: 52…53 °C/19 mmHg Melting Pt: 22.4 °C Density: 1.362 g/mL Flash Pt: 57 °C (135 °F) |
MDL Number:
MFCD00009715 CAS Number: 367-27-1 EINECS: 206-688-0 UN: 2921 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...