1,8-Diazabicyclo[5.4.0]undec-7-ene ≥99%

Supplier: BeanTown Chemical

217000-100G 217000-25G 217000-500G
BT217000-100GEA 715.28 USD
BT217000-100G BT217000-25G BT217000-500G
1,8-Diazabicyclo[5.4.0]undec-7-ene ≥99%
1,8-Diazabicyclo[5.4.0]undec-7-ene

CAS: 6674-22-2; EC No: 229-713-7; MDL No: MFCD00006930 UN No: UN3267; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C9H16N2; MW: 152.24 Boiling Point: 80-83°/0.6 mmHg; Flash point: 116°C (241°F) Density (g/mL): 1.018 Air Sensitive


  • Air Sensitive
  • Liquid

Formula: C₉H₁₆N₂
Boiling Pt: 80…83 °C/0.6 mmHg
Density: 1.018 g/mL
Flash Pt: 116 °C (241 °F)
MDL Number: MFCD00006930
CAS Number: 6674-22-2
EINECS: 229-713-7
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR