2-(Tributylstannyl)thiazole ≥96%
Supplier: BeanTown Chemical
216990-1G
216990-5G
BT216990-1GEA
132
USD
BT216990-1G
BT216990-5G
2-(Tributylstannyl)thiazole ≥96%
2-(Tributylstannyl)thiazole
CAS: 121359-48-6; MDL No: MFCD01319057
UN No: UN2788; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C15H29NSSn ; MW: 374.17
Boiling Point: 307-309°; Flash point: 110°C (230°F)
Density (g/mL): 1.19; Refractive Index: 1.520
- Refractive index: 1.520
- Liquid
Formula:
C₁₅H₂₉NSSn MW: 374.17 Boiling Pt: 307…309 °C Density: 1.19 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD01319057 CAS Number: 121359-48-6 ADR: 6.1,III |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...