L(-)-Camphor ≥98%

Supplier: BeanTown Chemical

Synonyms: (-)-CAMPHOR, (1S,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

216920-100G 216920-25G 216920-5G
BT216920-100GEA 596 USD
BT216920-100G BT216920-25G BT216920-5G
L(-)-Camphor ≥98%
L(-)-Camphor

CAS: 464-48-2; EC No: 207-354-7; MDL No: MFCD00064148; RTECS: EX1250000 UN No: UN2717; Haz Class: 4.1; Packing Group: III Crystalline; Molecular Formula: C10H16O; MW: 152.24 Boiling Point: 204°; Flash point: 65°C (149°F) Density (g/mL): 0.99; Optical Rotation: [α]20/D -43°, c = 10 in ethanol


  • RTECS: EX1250000
  • Optical rotation: [α]20/D -43°, c = 10 in ethanol
  • Crystalline

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C
Density: 0.99 g/mL
Flash Pt: 65 °C (149 °F)
MDL Number: MFCD00064148
CAS Number: 464-48-2
EINECS: 207-354-7
UN: 2717
ADR: 4.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR