Iridium(III) chloride, anhydrous ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
216825-1G
216825-5G
BT216825-1GEA
334
USD
BT216825-1G
BT216825-5G
Iridium(III) chloride, anhydrous ≥99.99% (trace metals basis)
Iridium(III) chloride
CAS: 10025-83-9; EC No: 233-044-6; MDL No: MFCD00011063
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: IrCl3; MW: 298.56
Melting Point: 763°
Hygroscopic
- Hygroscopic
- Powder
Formula:
Cl₃Ir MW: 298.58 g/mol Melting Pt: 763 °C |
MDL Number:
MFCD00011063 CAS Number: 10025-83-9 EINECS: 233-044-6 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...