Ammonium hydrogen difluoride ≥95% remainder mainly ammonium fluoride
Supplier: BeanTown Chemical
216795-100G
216795-2.5KG
216795-500G
BT216795-100GEA
34.61
USD
BT216795-100G
BT216795-2.5KG
BT216795-500G
Ammonium hydrogen difluoride ≥95% remainder mainly ammonium fluoride
Ammonium hydrogen difluoride
CAS: 1341-49-7; EC No: 215-676-4; MDL No: MFCD00012018; RTECS: BQ9200000
UN No: UN1727; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: (NH4)HF2 ; MW: 57.04
Melting Point: 125°
Hygroscopic
- RTECS: BQ9200000
- Hygroscopic
- Crystalline
Formula:
NH₄HF₂ MW: 57.04 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00012018 CAS Number: 1341-49-7 EINECS: 215-676-4 UN: 1727 ADR: 8,II |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...